Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TNF-alpha/TNFSF2 Hu 50ug
SDP

Supplier:  Medchemexpress LLC HYP705850UG

Encompass_Preferred

Recombinant human tumor necrosis factor alpha (TNF-α) for research use in studies of inflammation, cell signaling, and apoptosis. The protein corresponds to residues V77-L233 (157 amino acids) and is supplied lyophilized; follow manufacturer guidance for reconstitution and storage to preserve activity.

  • Recombinant human TNF-alpha corresponding to 157 amino acids (V77-L233).
  • Expressed in E. coli for research-grade production.
  • Supplied in multiple sizes, including a 50 μg vial.
  • Store at -20°C for up to 2 years from receipt.
  • After reconstitution, stable at 4°C for 1 week or at -20°C for longer with carrier protein.
  • Do not reconstitute below 100 μg/mL in ddH2O; freeze aliquots for extended storage.
  • Recommended stabilizers: 0.1% BSA, 5% HSA, 10% FBS, or 5% trehalose.

Catalog No. 50-001-84141


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.