Learn More
Medchemexpress LLC TNF-alpha/TNFSF2 Mo 100ug
SDP
Supplier:  Medchemexpress LLC HYP7090100UG
Encompass_Preferred
Recombinant mouse Tumor Necrosis Factor alpha (TNF-α/TNFSF2), 156 amino acids (L80-L235), supplied lyophilized for research use. Produced in E. coli and intended for in vitro studies of inflammation, cell signaling, and related assays. Follow manufacturer guidance for reconstitution and storage to preserve activity.
  • Biologically active cytokine for inflammation and signaling studies.
  • Produced in E. coli and purified to >95% by reducing SDS-PAGE.
  • Approximate molecular weight 17.3 kDa based on SDS-PAGE.
  • Lyophilized from PBS, pH 7.4, with optional 8% trehalose for stability.
  • Stable when stored at -20°C; aliquot and freeze at -20°C or -80°C for extended storage.
  • Available in multiple sizes to suit experimental needs.
Catalog No. 50-001-97271