Learn More
Medchemexpress LLC TNF-alpha/TNFSF2 Mo 100ug
SDP
Catalog No. 5000197269
Encompass_Preferred
Recombinant mouse Tumour Necrosis Factor alpha (TNF-α) is a 156-amino-acid pro-inflammatory cytokine supplied as a lyophilized recombinant protein expressed in E. coli for research applications. It is used in studies of inflammation, apoptosis, and immune signaling and should be reconstituted and stored according to the product datasheet to preserve activity.
  • Produced in E. coli and provided as lyophilized powder.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Approximate molecular weight ~17.3 kDa under reducing conditions.
  • Sequence corresponds to L80-L235, 156 amino acids.
  • Supplied size 100 μg with reconstitution guidance to ≥100 μg/mL.
  • Stable when stored frozen; aliquot to avoid freeze-thaw cycles.
Catalog No. 50-001-97269 Supplier Medchemexpress LLC Supplier No. HYP7090A100UG
Recombinant mouse Tumour Necrosis Factor alpha (TNF-α) is a 156-amino-acid pro-inflammatory cytokine supplied as a lyophilized recombinant protein expressed in E. coli for research applications. It is used in studies of inflammation, apoptosis, and immune signaling and should be reconstituted and stored according to the product datasheet to preserve activity.
  • Produced in E. coli and provided as lyophilized powder.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Approximate molecular weight ~17.3 kDa under reducing conditions.
  • Sequence corresponds to L80-L235, 156 amino acids.
  • Supplied size 100 μg with reconstitution guidance to ≥100 μg/mL.
  • Stable when stored frozen; aliquot to avoid freeze-thaw cycles.