Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TNF-alpha/TNFSF2 Mo 100ug
SDP

Catalog No. 5000184793
Encompass_Preferred

Recombinant mouse TNF-alpha (amino acids D89-L235; 147 aa) supplied as a 100 μg lyophilized powder for research use. The protein is produced in E. coli, provided at >95% purity, and characterized for bioactivity in cytotoxicity assays; recommended storage and reconstitution guidance is included with the product.

  • Recombinant mouse TNF-alpha fragment (147 amino acids D89-L235).
  • Produced in E. coli expression system.
  • Supplied as a lyophilized powder for stability.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Predicted molecular mass approximately 16.4 kDa.
  • Bioactive in L-929 cytotoxicity assay; ED50 1.616 pg/mL.
  • Store at -20°C; aliquot to avoid repeated freeze-thaw.

Catalog No. 50-001-84793 Supplier Medchemexpress LLC Supplier No. HYP70571100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant mouse TNF-alpha (amino acids D89-L235; 147 aa) supplied as a 100 μg lyophilized powder for research use. The protein is produced in E. coli, provided at >95% purity, and characterized for bioactivity in cytotoxicity assays; recommended storage and reconstitution guidance is included with the product.

  • Recombinant mouse TNF-alpha fragment (147 amino acids D89-L235).
  • Produced in E. coli expression system.
  • Supplied as a lyophilized powder for stability.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Predicted molecular mass approximately 16.4 kDa.
  • Bioactive in L-929 cytotoxicity assay; ED50 1.616 pg/mL.
  • Store at -20°C; aliquot to avoid repeated freeze-thaw.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.