Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TNF-alpha/TNFSF2 Mo 1mg
SDP

Catalog No. 5000185520
Encompass_Preferred

Recombinant mouse TNF-alpha/TNFSF2 protein (147 amino acids, residues D89-L235) expressed in E. coli and supplied as a lyophilized powder for research applications. The product offers high purity and low endotoxin to support biochemical and cell-based assays, receptor binding studies, and signaling experiments.

  • Recombinant mouse TNF-alpha corresponding to residues D89-L235.
  • Produced in E. coli for consistent yield and quality.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Endotoxin level below 1 EU/μg by LAL assay.
  • Lyophilized formulation for convenient storage and transport.
  • Stable at -20°C for long-term storage; aliquot after reconstitution.

Catalog No. 50-001-85520 Supplier Medchemexpress LLC Supplier No. HYP705711MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse TNF-alpha/TNFSF2 protein (147 amino acids, residues D89-L235) expressed in E. coli and supplied as a lyophilized powder for research applications. The product offers high purity and low endotoxin to support biochemical and cell-based assays, receptor binding studies, and signaling experiments.

  • Recombinant mouse TNF-alpha corresponding to residues D89-L235.
  • Produced in E. coli for consistent yield and quality.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Endotoxin level below 1 EU/μg by LAL assay.
  • Lyophilized formulation for convenient storage and transport.
  • Stable at -20°C for long-term storage; aliquot after reconstitution.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.