Learn More
Medchemexpress LLC TNF-alpha/TNFSF2 Mo 2ug
SDP
Supplier:  Medchemexpress LLC HYP70902UG
Encompass_Preferred
Recombinant mouse Tumour Necrosis Factor alpha (TNF-alpha, TNFSF2) is supplied as the mature 156-amino-acid fragment (residues L80-L235) produced in Escherichia coli for research applications. The protein engages TNF receptors and activates downstream signaling pathways involved in inflammation, apoptosis, cancer biology, and neurogenesis.
  • Mature 156-amino-acid fragment corresponding to residues L80-L235.
  • Expressed in Escherichia coli for reproducible production.
  • Activates TNFR1 and TNFR2 signaling pathways.
  • Suitable for studies of inflammation, apoptosis, and cancer biology.
  • Provided for research use; follow manufacturer guidance for handling and storage.
Catalog No. 50-001-97143