Learn More
Medchemexpress LLC TNF-alpha/TNFSF2 Mo 2ug
SDP
Catalog No. 5000184149
Encompass_Preferred
Recombinant mouse TNF-alpha/TNFSF2 protein (147 aa), corresponding to residues D89-L235 of mouse TNF-α, expressed in E. coli and supplied lyophilized. The product is >95% pure by reducing SDS-PAGE, has an apparent molecular weight of ~17.0 kDa, and is functionally validated in an L-929 cytotoxicity assay (ED50 = 1.616 pg/mL), making it suitable for signaling, bioactivity, and cytokine functional studies.
  • Produced in E. coli for efficient bacterial expression.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Corresponds to amino acids D89-L235 of mouse TNF-α (147 aa).
  • Apparent molecular weight of approximately 17.0 kDa.
  • Validated bioactivity in L-929 cytotoxicity assay (ED50 = 1.616 pg/mL).
  • Supplied lyophilized for long-term storage and easy reconstitution.
Catalog No. 50-001-84149 Supplier Medchemexpress LLC Supplier No. HYP705712UG
Recombinant mouse TNF-alpha/TNFSF2 protein (147 aa), corresponding to residues D89-L235 of mouse TNF-α, expressed in E. coli and supplied lyophilized. The product is >95% pure by reducing SDS-PAGE, has an apparent molecular weight of ~17.0 kDa, and is functionally validated in an L-929 cytotoxicity assay (ED50 = 1.616 pg/mL), making it suitable for signaling, bioactivity, and cytokine functional studies.
  • Produced in E. coli for efficient bacterial expression.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Corresponds to amino acids D89-L235 of mouse TNF-α (147 aa).
  • Apparent molecular weight of approximately 17.0 kDa.
  • Validated bioactivity in L-929 cytotoxicity assay (ED50 = 1.616 pg/mL).
  • Supplied lyophilized for long-term storage and easy reconstitution.