Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TNF-alpha/TNFSF2 Mo 50ug
SDP

Catalog No. 5000184621
Encompass_Preferred

Recombinant mouse Tumor Necrosis Factor alpha (TNF-alpha/TNFSF2) is supplied as a lyophilized protein corresponding to the C-terminal extracellular domain (D89-L235, 147 aa). Expressed in E. coli and purified to >95% by reducing SDS-PAGE, the protein is provided in multiple sizes (including 50 μg) for use in studies of inflammation, cell signaling, and immune response modulation.

  • Lyophilized recombinant protein, 147 amino acids (D89-L235).
  • Expressed in E. coli for consistent production.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Molecular weight approximately 17.0 kDa under reducing conditions.
  • Available in multiple sizes, including a 50 μg option.
  • Stable when stored at -20°C; aliquot and store at -80°C for long-term use.
  • Reconstitute at a minimum concentration of 100 μg/mL in ddH2O; add carrier protein for long-term storage.

Catalog No. 50-001-84621 Supplier Medchemexpress LLC Supplier No. HYP7057150UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse Tumor Necrosis Factor alpha (TNF-alpha/TNFSF2) is supplied as a lyophilized protein corresponding to the C-terminal extracellular domain (D89-L235, 147 aa). Expressed in E. coli and purified to >95% by reducing SDS-PAGE, the protein is provided in multiple sizes (including 50 μg) for use in studies of inflammation, cell signaling, and immune response modulation.

  • Lyophilized recombinant protein, 147 amino acids (D89-L235).
  • Expressed in E. coli for consistent production.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Molecular weight approximately 17.0 kDa under reducing conditions.
  • Available in multiple sizes, including a 50 μg option.
  • Stable when stored at -20°C; aliquot and store at -80°C for long-term use.
  • Reconstitute at a minimum concentration of 100 μg/mL in ddH2O; add carrier protein for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.