Learn More
Medchemexpress LLC TNF-alpha/TNFSF2 Mo 50ug
SDP
Catalog No. 5000185367
Encompass_Preferred
Recombinant mouse TNF-alpha (156 a.a) supplied lyophilized for use in in vitro functional and biochemical studies. Expressed in E. coli, the protein is highly pure and validated for activity in cytotoxicity and signaling assays.
  • Lyophilized formulation for long-term storage and easy reconstitution.
  • High purity suitable for biochemical and cell-based assays.
  • Low endotoxin level (<0.2 EU/μg) for sensitive applications.
  • Validated biological activity (ED50 <0.1 ng/mL in L-929 cell assay).
  • Multiple pack sizes available to accommodate experimental scale.
Catalog No. 50-001-85367 Supplier Medchemexpress LLC Supplier No. HYP7090A50UG
Recombinant mouse TNF-alpha (156 a.a) supplied lyophilized for use in in vitro functional and biochemical studies. Expressed in E. coli, the protein is highly pure and validated for activity in cytotoxicity and signaling assays.
  • Lyophilized formulation for long-term storage and easy reconstitution.
  • High purity suitable for biochemical and cell-based assays.
  • Low endotoxin level (<0.2 EU/μg) for sensitive applications.
  • Validated biological activity (ED50 <0.1 ng/mL in L-929 cell assay).
  • Multiple pack sizes available to accommodate experimental scale.