Learn More
Medchemexpress LLC TNF-alpha/TNFSF2 Mo 50ug
SDP
Supplier:  Medchemexpress LLC HYP709050UG
Encompass_Preferred
Recombinant mouse TNF-alpha (TNFSF2) is a pro-inflammatory cytokine supplied as a purified recombinant protein for research use. Supplied as a defined 50 μg quantity, it is intended for in vitro studies of inflammation, apoptosis, and cytokine-mediated signaling.
  • Suitable for in vitro cell signaling and cytokine assays.
  • Recombinant protein corresponding to 156 amino acids.
  • Offered in a small, convenient quantity for focused experiments.
  • Intended for research use only, not for clinical or diagnostic use.
Catalog No. 50-001-84144