Learn More
Medchemexpress LLC TNF-alpha/TNFSF2 Mo 50ug
SDP
Catalog No. 5000179654
Encompass_Preferred
TNF-alpha/TNFSF2 Protein Mouse (His) is recombinant mouse tumor necrosis factor alpha (TNF- ) with his tag TNF-alpha/TNFSF2 Protein a major mediator of inflammation is involved in a number of infectious and parasitic diseases
Catalog No. 50-001-79654 Supplier Medchemexpress LLC Supplier No. HYP741750UG
TNF-alpha/TNFSF2 Protein Mouse (His) is recombinant mouse tumor necrosis factor alpha (TNF- ) with his tag TNF-alpha/TNFSF2 Protein a major mediator of inflammation is involved in a number of infectious and parasitic diseases