Learn More
Medchemexpress LLC TNF-alpha/TNFSF2 Mo 5ug
SDP
Catalog No. 5000197091
Encompass_Preferred
TNF-alpha/TNFSF2 Protein, Mouse is a recombinant murine tumor necrosis factor alpha comprising the extracellular domain (156 amino acids). Supplied lyophilized and produced in E. coli, the protein is intended for in vitro research to support studies of inflammatory signaling through TNFR1/TNFR2 and downstream pathways such as NF-κB and JNK.
Catalog No. 50-001-97091 Supplier Medchemexpress LLC Supplier No. HYP70905UG
TNF-alpha/TNFSF2 Protein, Mouse is a recombinant murine tumor necrosis factor alpha comprising the extracellular domain (156 amino acids). Supplied lyophilized and produced in E. coli, the protein is intended for in vitro research to support studies of inflammatory signaling through TNFR1/TNFR2 and downstream pathways such as NF-κB and JNK.