Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TNF-alpha/TNFSF2 Ra 10ug
SDP

Catalog No. 5000184223
Encompass_Preferred

Recombinant rat Tumor Necrosis Factor-alpha (TNF-α) is a proinflammatory cytokine supplied lyophilized for use in cellular and biochemical research. The product comprises the C-terminal fragment (residues L80-L235), expressed recombinantly and purified for studies of inflammation, apoptosis, and TNF-mediated signaling.

  • Purity greater than 95% by reducing SDS-PAGE.
  • Expressed in Escherichia coli for recombinant production.
  • Lyophilized from 0.2 μm filtered PBS, pH 7.4.
  • Predicted molecular mass ~17.4 kDa (observed 14-17 kDa).
  • Storage recommended at -20°C with aliquots for long-term -80°C storage.
  • Suitable for in vitro studies of inflammation and signaling pathways.

Catalog No. 50-001-84223 Supplier Medchemexpress LLC Supplier No. HYP7069710UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rat Tumor Necrosis Factor-alpha (TNF-α) is a proinflammatory cytokine supplied lyophilized for use in cellular and biochemical research. The product comprises the C-terminal fragment (residues L80-L235), expressed recombinantly and purified for studies of inflammation, apoptosis, and TNF-mediated signaling.

  • Purity greater than 95% by reducing SDS-PAGE.
  • Expressed in Escherichia coli for recombinant production.
  • Lyophilized from 0.2 μm filtered PBS, pH 7.4.
  • Predicted molecular mass ~17.4 kDa (observed 14-17 kDa).
  • Storage recommended at -20°C with aliquots for long-term -80°C storage.
  • Suitable for in vitro studies of inflammation and signaling pathways.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.