Learn More
Medchemexpress LLC HY-P7304 10ug Medchemexpress, TNF-beta, Human CAS: Purity:>98%
SDP
Catalog No. 502083143
Encompass_Preferred
Medchemexpress, HY-P7304 10ug TNF-beta, Human CAS: Recombinant Human Tumor Necrosis Factor beta exhibits anti-proliferative activity and causes the cellular destruction of tumor cell lines and performs a variety of important roles in immune regulation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3143 Supplier Medchemexpress LLC Supplier No. HYP730410UG
Medchemexpress, HY-P7304 10ug TNF-beta, Human CAS: Recombinant Human Tumor Necrosis Factor beta exhibits anti-proliferative activity and causes the cellular destruction of tumor cell lines and performs a variety of important roles in immune regulation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.