Learn More
AdipoGen TNF-R1 (human):Fc (human) (rec.) (50 µg)
SDP
Supplier:  AdipoGen AG40B0074C050
Encompass_Preferred
Protein. The extracellular domain of human TNF-R1 (aa 22-211) is fused to the Fc portion of human IgG1. Source: HEK 293 cells. Endotoxin content: <0.01EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains PBS. Binds human and mouse TNF-alpha. Purity: >95% (SDS-PAGE). TNF-R1 is a receptor for TNF-alpha and homotrimeric lymphotoxin-alpha. The adapter molecule FADD recruits caspase-8 to the activated receptor. The resulting death-inducing signaling complex (DISC) performs caspase-8 proteolytic activation which initiates the subsequent cascade of caspases (aspartate-specific cysteine proteases) mediating apoptosis.
Catalog No. 50-205-3131