Learn More
Enzo Life Sciences TNF-R1 (human) monoclonal antibody (H398) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804200C100
Encompass_Preferred
Clone: H398. Isotype: IgG2a. Immunogen: Human TNF-R1. Species reactivity: Human. Specificity: Recognizes membrane-bound and soluble TNF-R1. Applications: Flow Cytometry, IHC (FS), FUNC. Purity: Protein G affinity purified. Formulation: Liquid. In PBS.
Catalog No. 50-200-9823