Learn More
Enzo Life Sciences TNF-R2 (human) : Fc (human), (recombinant) (50µg)
SDP
Supplier:  Enzo Life Sciences ALX522014C050
Encompass_Preferred
Alternative name: p75, CD120b, TNF-R 80kDa, TNFRSF 1B. Concentration: 1mg/ml after reconstitution. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized. Contains PBS. Source: Produced in HEK 293 cells. The extracellular domain of human TNF-R2 (aa 1-257) is fused to the Fc portion of human IgG1. Specificity: Binds human and mouse TNF-α (tumor necrosis factor-α). Long Term Storage: -20°C. Applications: ELISA .
Catalog No. 50-200-9364