Learn More
Enzo Life Sciences TNF-R2 monoclonal antibody (80M2) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804450C100
Encompass_Preferred
Clone: 80M2. Isotype: IgG1. Immunogen: Human TNF-R2. Species reactivity: Human, Rat. Specificity: Recognizes the extracellular part of human and rat TNF-R2 and soluble human TNF-R2. Applications: ELISA, Flow Cytometry, IHC (FS), IP, WB, FUNC. Formulation: Liquid. 0.2µm-filtered solution in PBS containing 0.1% BSA.
Catalog No. 50-200-9912