Learn More
Enzo Life Sciences TNF receptor 1 polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADICSA815F
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of mouse TNF-R1. The sequence is completely conserved in rat and human. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Yeast, Xenopus, Sheep, Rabbit, Porcine, Monkey, Hamster, Drosophila, Dog, Bovine. Applications: IHC (PS), IP, WB. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7200