Learn More
Medchemexpress LLC TNF RI/TNFRSF1A Mou 10ug
SDP
Supplier:  Medchemexpress LLC HYP7344910UG
Encompass_Preferred
TNFRSF1A (TNF RI) protein has a high abilityto bind with tumor necrosis factor-alpha (TNF- ) TNFRSF1A is a STAT3 targetgene that regulates the NF- B pathway TNFRSF1A activate NF- B mediateapoptosis and function as a regulator of inflammation TNF RI/TNFRSF1A Protein Mouse (HEK293 Fc) is expressed by HEK 293 cells and has a transmembrane region with a Fc tag at the C-terminus[1]
Catalog No. 50-001-80550