Learn More
Medchemexpress LLC TNF RI/TNFRSF1A Mou 10ug
SDP
Supplier:  Medchemexpress LLC HYP7345010UG
Encompass_Preferred
TNFRSF1A (TNF RI) protein has a highability to bind with tumor necrosis factor-alpha (TNF- ) TNFRSF1A is a STAT3target gene that regulates the NF- B pathway TNFRSF1A activate NF-kappaB mediate apoptosis and function as a regulator of inflammation TNF RI/TNFRSF1AProtein Mouse (HEK293 His) is expressed by HEK 293 cells and has atransmembrane region with a His tag at theC-terminus[1]
Catalog No. 50-001-80646