Learn More
Medchemexpress LLC TNF RI/TNFRSF1A Mou 10ug
SDP
Catalog No. 5000183120
Encompass_Preferred
TNFRSF1A (TNF RI) protein has a highability to bind with tumor necrosis factor-alpha (TNF- ) TNFRSF1A is a STAT3target gene that regulates the NF- B pathway TNFRSF1A activate NF- B mediateapoptosis and function as a regulator of inflammation TNF RI/TNFRSF1AProtein Mouse is expressed by E coli and has atransmembrane region (I22-A212)[1]
Catalog No. 50-001-83120 Supplier Medchemexpress LLC Supplier No. HYP7244310UG
TNFRSF1A (TNF RI) protein has a highability to bind with tumor necrosis factor-alpha (TNF- ) TNFRSF1A is a STAT3target gene that regulates the NF- B pathway TNFRSF1A activate NF- B mediateapoptosis and function as a regulator of inflammation TNF RI/TNFRSF1AProtein Mouse is expressed by E coli and has atransmembrane region (I22-A212)[1]