Learn More
Medchemexpress LLC TNF RII/TNFRSF1B Cy 10ug
SDP
Catalog No. 5000183123
Encompass_Preferred
TNFRII (TNFRSF1B) protein has a high ability to bind with tumor necrosis factor-alpha (TNF- ) TNFRII has pro-apoptotic function TNFRII recruits TRAF2 induces gene expression and intensively crosstalks with TNF-R1 TNFRII selectively enhances the induction of apoptosis by the death receptor TNFRI TNF RII/TNFRSF1B Protein Cynomolgus/Rhesus Macaque (HEK293 His) is expressed by HEK293 with 6 His tag at the C-terminus[1]
Catalog No. 50-001-83123 Supplier Medchemexpress LLC Supplier No. HYP7244410UG
TNFRII (TNFRSF1B) protein has a high ability to bind with tumor necrosis factor-alpha (TNF- ) TNFRII has pro-apoptotic function TNFRII recruits TRAF2 induces gene expression and intensively crosstalks with TNF-R1 TNFRII selectively enhances the induction of apoptosis by the death receptor TNFRI TNF RII/TNFRSF1B Protein Cynomolgus/Rhesus Macaque (HEK293 His) is expressed by HEK293 with 6 His tag at the C-terminus[1]