Learn More
Medchemexpress LLC TNF RII/TNFRSF1B Mo 10ug
SDP
Catalog No. 5000185149
Encompass_Preferred
MedChemExpress recombinant mouse TNF RII (TNFRSF1B) extracellular domain expressed in HEK293 cells with a C-terminal 6xHis tag. This recombinant receptor fragment is supplied for research use in biochemical and cell-based studies, and is suitable for binding assays, signaling experiments, and assay development.
  • Expressed in HEK293 mammalian cells for correct folding and glycosylation.
  • Contains a C-terminal 6xHis tag for purification and detection.
  • Provides a biologically relevant receptor extracellular domain for binding studies.
  • Supplied at research-appropriate quantities for assay development.
  • Suitable for use in biochemical assays and cell-based signaling experiments.
  • Quality controlled for purity and activity where applicable.
Catalog No. 50-001-85149 Supplier Medchemexpress LLC Supplier No. HYP7051610UG
MedChemExpress recombinant mouse TNF RII (TNFRSF1B) extracellular domain expressed in HEK293 cells with a C-terminal 6xHis tag. This recombinant receptor fragment is supplied for research use in biochemical and cell-based studies, and is suitable for binding assays, signaling experiments, and assay development.
  • Expressed in HEK293 mammalian cells for correct folding and glycosylation.
  • Contains a C-terminal 6xHis tag for purification and detection.
  • Provides a biologically relevant receptor extracellular domain for binding studies.
  • Supplied at research-appropriate quantities for assay development.
  • Suitable for use in biochemical assays and cell-based signaling experiments.
  • Quality controlled for purity and activity where applicable.