Learn More
Medchemexpress LLC TNF RII/TNFRSF1B Mo 2ug
SDP
Supplier:  Medchemexpress LLC HYP705162UG
Encompass_Preferred
Recombinant mouse TNF RII (TNFRSF1B) extracellular domain produced in HEK293 cells with a C-terminal His tag. Designed for biochemical and cell-based research to study TNF-α binding, signaling, and apoptosis pathways. Supplied as a purified recombinant protein for research use.
  • Recombinant extracellular domain of mouse TNFRII.
  • Expressed in HEK293 cells for mammalian post-translational modifications.
  • C-terminal hexahistidine tag enables purification and detection.
  • Suitable for binding assays and signaling studies involving TNF-α.
  • Intended for research use; not for therapeutic or diagnostic applications.
Catalog No. 50-001-84654