Learn More
Cayman Chemical TNFa ChImRIc MouseHumn Mo
SDP
Supplier:  Cayman Chemical 37178200 UG
Encompass_Preferred
Immunogen: Human TNF-α • Host: Chimeric Monoclonal Antibody • Species Reactivity: (+) Human, rat • Cross Reactivity: (+) TNF-α • Applications: ELISA
Catalog No. 50-291-9456