Learn More
Cayman Chemical TNFa ChImRIc MouseHumn Mo
SDP
Catalog No. 502919392
Encompass_Preferred
Immunogen: Human TNF-α • Host: Chimeric Monoclonal Antibody • Species Reactivity: (+) Human • Cross Reactivity: (+) TNF-α, soluble TNF-α; (-) TNF-β • Applications: ELISA, IF, IHC
Catalog No. 50-291-9392 Supplier Cayman Chemical Supplier No. 37180200 UG
Immunogen: Human TNF-α • Host: Chimeric Monoclonal Antibody • Species Reactivity: (+) Human • Cross Reactivity: (+) TNF-α, soluble TNF-α; (-) TNF-β • Applications: ELISA, IF, IHC