Learn More
Cayman Chemical TNFa ChImRIc MouseMouse Mo
SDP
Catalog No. 502919455
Encompass_Preferred
Immunogen: Human TNF-α • Host: Chimeric Monoclonal Antibody • Species Reactivity: (+) Human • Cross Reactivity: (+) TNF-α • Applications: ELISA
Catalog No. 50-291-9455 Supplier Cayman Chemical Supplier No. 37177200 UG
Immunogen: Human TNF-α • Host: Chimeric Monoclonal Antibody • Species Reactivity: (+) Human • Cross Reactivity: (+) TNF-α • Applications: ELISA