Learn More
Cayman Chemical TNFa ChImRIc MousRabbIt M
SDP
Supplier:  Cayman Chemical 37182200 UG
Encompass_Preferred
Immunogen: Human TNF-α • Host: Chimeric Monoclonal Antibody • Species Reactivity: (+) Human • Cross Reactivity: (+) TNF-α, soluble TNF-α; (-) TNF-β • Applications: ELISA, IF, IHC
Catalog No. 50-291-9350