Learn More
Cayman Chemical TNFa ChImRIc MousRabbIt M
SDP
Catalog No. 502919391
Encompass_Preferred
Immunogen: Human TNF-α • Host: Chimeric Monoclonal Antibody • Species Reactivity: (+) Human • Cross Reactivity: (+) TNF-α • Applications: ELISA
Catalog No. 50-291-9391 Supplier Cayman Chemical Supplier No. 37179200 UG
Immunogen: Human TNF-α • Host: Chimeric Monoclonal Antibody • Species Reactivity: (+) Human • Cross Reactivity: (+) TNF-α • Applications: ELISA