Learn More
Enzo Life Sciences TNF-alpha Recombinant monoclonal antibody (CDP 571) (Rabbit IgG kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS5630200
Encompass_Preferred
Clone: CDP 571 (Humicade). Isotype: IgGκ. Immunogen: Human TNF-α. Species reactivity: Human. Applications: ELISA. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3089