Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 7124MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: P/T2. Clonality: Monoclonal. Host: Mouse. Isotype: IgM kappa. Positive Control: HeLa, HL-60, or A431 cells. Macrophages in lymph node or Tonsil. Cellular Staining: Cytoplasmic, extracellular (secreted). Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: A hexadecapeptide corresponding to aa115-130 (NGVELRDNQLVVPSEG) of human TNF-?, conjugated to thyroglobulin. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 17kDa.Chromosome: 6p21.33. Gene Symbol: TNF. Gene ID: 7124. Ensemble: ENSG00000204490. Uniprot: NP_000585.
Catalog No. 50-188-5229