Learn More
Sino Biological TNF-alpha / TNFA / TNFSF2 Antibody, Mouse MAb 100uL
Catalog No. 502248070
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TNF-alpha / TNFA / TNFSF2 (rh TNF-alpha / TNFA / TNFSF2; Catalog#10602-HNAE; NP_000585.2; Val77-Leu233). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
This item is not returnable. View return policy
Catalog No. 50-224-8070 Supplier Sino Biological Supplier No. 10602MM08100
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TNF-alpha / TNFA / TNFSF2 (rh TNF-alpha / TNFA / TNFSF2; Catalog#10602-HNAE; NP_000585.2; Val77-Leu233). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.