Learn More
Sino Biological TNF-alpha / TNFA / TNFSF2 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Catalog No. 502248010
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human TNF-alpha / TNFA (rh TNF-alpha / TNFA; Catalog#10602-HNAE; NP_000585.2; Val77-Leu233). TNF-alpha / TNFA specific IgG was purified by Human TNF-alpha / TNFA affinity chromatography.
This item is not returnable. View return policy
Catalog No. 50-224-8010 Supplier Sino Biological Supplier No. 10602T16100
Produced in rabbits immunized with purified, recombinant Human TNF-alpha / TNFA (rh TNF-alpha / TNFA; Catalog#10602-HNAE; NP_000585.2; Val77-Leu233). TNF-alpha / TNFA specific IgG was purified by Human TNF-alpha / TNFA affinity chromatography.