Learn More
Sino Biological TNF-alpha / TNFA / TNFSF2 Neutralizing Antibody 500ug
Catalog No. 502247753
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human TNF-alpha / TNFA (rHuman TNF-alpha / TNFA; Catalog#10602-HNAE; NP_000585.2; Val77-Leu233) and was produced using recombinant antibody technology.
This item is not returnable. View return policy
Catalog No. 50-224-7753 Supplier Sino Biological Supplier No. 10602R10N1500
This antibody was obtained from a rabbit immunized with purified, recombinant Human TNF-alpha / TNFA (rHuman TNF-alpha / TNFA; Catalog#10602-HNAE; NP_000585.2; Val77-Leu233) and was produced using recombinant antibody technology.