| Accession Number |
P25118 |
| Assay Range |
1.23-300 pg/mL |
| Assay Sensitivity |
1 pg/mL |
| Calibration Range |
1 pg/mL to 300 pg/mL |
| Assay |
Solid-phase sandwich ELISA |
| Conjugate |
HRP |
| Immunoassay Kit Format |
Sandwich ELISA Kit |
| Product Type |
ELISA |
| Protein Family |
Cytokines & Receptors |
| Sample Type |
Plasma, Serum, Supernatant |
| Specificity |
This ELISA pair antibody detects mouse sTNFRI. Other species not determined. |
| Sufficient For |
96 Assays |
| Target Species |
Mouse |
| For Use With (Application) |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis |
| For Use With (Equipment) |
Absorbance Microplate Reader |
| Gene Alias |
CD120a; CD120a antigen; FPF; hypothetical protein; metalloproteinase inhibitor 4; MGC19588; MS5; n-smase activation assoc. factor; p55; p55 TNF receptor; p55-R; p60; p62; p63; sCD120a; soluble CD120a; soluble TNFR1; sTNF RI; sTNFR I; sTNFR1; sTNFRI; TBP1; TBPI; TIMP metallopeptidase inhibitor 4; Timp4; TIMP-4; tissue inhibitor of metalloproteinase 4; tissue inhibitor of metalloproteinases 4; TNF receptor alpha chain; TNF receptor superfamily member 1A; TNF-alphaR1; TNF-alpha-R1; TNFalpha-R1; TNFAR; TNFR; TNF-R; TNFR I; TNFR1; TNF-R1; Tnfr-1; TNFR1-d2; Tnfr-2; TNFR55; TNF-R55; TNFR60; TNFRI; TNF-RI; TNF-R-I; TNFR-I; TNFRp55; Tnfrsf1a; tumor necrosis factor binding protein 1; Tumor necrosis factor receptor; tumor necrosis factor receptor 1; tumor necrosis factor receptor 1A isoform beta; tumor necrosis factor receptor superfamily member 1A; Tumor necrosis factor receptor superfamily member 1A, membrane form; tumor necrosis factor receptor superfamily, member 1A; tumor necrosis factor receptor type 1; tumor necrosis factor receptor type I; tumor necrosis factor-alpha receptor; Tumor necrosis factor-binding protein 1CD120a; CD120a antigen; FPF; hypothetical protein; metalloproteinase inhibitor 4; MGC19588; MS5; n-smase activation assoc. factor; p55; p55 TNF receptor; p55-R; p60; p62; p63; sCD120a; soluble CD120a; soluble TNFR1; sTNF RI; sTNFR I; sTNFR1; sTNFRI; TBP1; TBPI; TIMP metallopeptidase inhibitor 4; Timp4; TIMP-4; tissue inhibitor of metalloproteinase 4; tissue inhibitor of metalloproteinases 4; TNF receptor alpha chain; TNF receptor superfamily member 1A; TNF-alphaR1; TNF-alpha-R1; TNFalpha-R1; TNFAR; TNFR; TNF-R; TNFR I; TNFR1; TNF-R1; Tnfr-1; TNFR1-d2; Tnfr-2; TNFR55; TNF-R55; TNFR60; TNFRI; TNF-RI; TNF-R-I; TNFR-I; TNFRp55; Tnfrsf1a; tumor necrosis factor binding protein 1; Tumor necrosis factor receptor; tumor necrosis factor receptor 1; tumor necrosis factor receptor 1A isoform beta; tumor necrosis factor receptor superfamily member 1A; Tumor necrosis factor receptor superfamily member 1A, membrane form; tumor necrosis factor receptor superfamily, member 1A; tumor necrosis factor receptor type 1; tumor necrosis factor receptor type I; tumor necrosis factor-alpha receptor; Tumor necrosis factor-binding protein 1; CD120a; FPF; TNF-R; TNF-R-I; TNF-R1; TNF-R55; TNF-alphaR1; TNFAR; TNFR60; TNFRI; TNFRp55; TNFalpha-R1; Tnfr-2; Tnfr1; p55; p55-R |
| Gene ID (Entrez) |
21937 |
| Gene Symbol |
Tnfrsf1a |
| Interassay CV |
<12% |
| Intraassay CV |
<10% |
| Kit Contents |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers |
| Label or Dye |
Biotin |
| Quantity |
10 x 96 Tests |
| Regulatory Status |
RUO |
| Research Discipline |
Inflammation |
| Sample Volume |
Plasma 20 μL, Serum 20 μL, Supernatant 50 μL |
| Shipping Condition |
Wet or Dry Ice |
| Storage Requirements |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C |
| Synonym |
CD120a, FPF, TNF-R, TNF-R-I, TNF-R1, TNF-R55, TNF-alphaR1, TNFAR, TNFR60, TNFRI, TNFRp55, TNFalpha-R1, Tnfr-2, Tnfr1, p55, p55-R |
| Target |
TNFR1 |
| Test Time |
1 hr 20 min |
| Total Assay Time |
4 hr 45 min |
|
Show More
Show Less
|
|