Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Human TNFR1 Protein, Fc Tag, Lyophilized, 100 μg

Catalog No. 501658101
Encompass_Preferred

Human TNFR1, also known as TNFRSF1A or CD120a, is a member of the tumor necrosis factor receptor superfamily that functions as a receptor for TNFSF2 and TNFSF1. This protein is expressed in human 293 cells and carries a human IgG1 Fc tag at the C-terminus. It is lyophilized from a 0.22 μm filtered solution containing Tris, glycine, arginine, and NaCl with trehalose as a protectant, and is MALS-verified for quality assurance.

  • Greater than 95% purity as determined by SDS-PAGE
  • Calculated molecular weight of 47.1 kDa
  • Endotoxin level of less than 1.0 EU per μg
  • Expressed from human HEK293 cells
  • Contains amino acids Leu 30 to Thr 211

Catalog No. 50-165-8101 Supplier ACROBiosystems Supplier No. TN1H5251100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Human TNFR1, also known as TNFRSF1A or CD120a, is a member of the tumor necrosis factor receptor superfamily that functions as a receptor for TNFSF2 and TNFSF1. This protein is expressed in human 293 cells and carries a human IgG1 Fc tag at the C-terminus. It is lyophilized from a 0.22 μm filtered solution containing Tris, glycine, arginine, and NaCl with trehalose as a protectant, and is MALS-verified for quality assurance.

  • Greater than 95% purity as determined by SDS-PAGE
  • Calculated molecular weight of 47.1 kDa
  • Endotoxin level of less than 1.0 EU per μg
  • Expressed from human HEK293 cells
  • Contains amino acids Leu 30 to Thr 211

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.