Learn More
ACROBiosystems Recombinant Protein;Human TNFR1 / CD120a / TNFRSF1A Protein;HEK293;100UG;TN1,TNFR1,CD120a,TNFRSF1A,FPF,TBP1,TNF-R,TNF-R-I,TNF-R55,TNFAR,TNFR-1,TNFR55,TNFR60,p55,p60
Catalog No. 501658095
Encompass_Preferred
Recombinant Protein;Human TNFR1 / CD120a / TNFRSF1A Protein;100UG;Human TNFR1, His Tag (TN1-H5222) is expressed from human 293 cells (HEK293). It contains AA Ile 22 - Thr 211 (Accession # NP_001056).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.0 kDa. The protein migrates as 28-38 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.;Immobilized Human TNF-alpha, Tag Free (Cat. No. TNA-H4211) at 2 μg/mL (100 μL/well) can bind Human TNFR1, His Tag (Cat. No. TN1-H5222) with a linear range of 3-25 ng/mL (QC tested).
Catalog No. 50-165-8095 Supplier ACROBiosystems Supplier No. TN1H5222100UG
Recombinant Protein;Human TNFR1 / CD120a / TNFRSF1A Protein;100UG;Human TNFR1, His Tag (TN1-H5222) is expressed from human 293 cells (HEK293). It contains AA Ile 22 - Thr 211 (Accession # NP_001056).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.0 kDa. The protein migrates as 28-38 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.;Immobilized Human TNF-alpha, Tag Free (Cat. No. TNA-H4211) at 2 μg/mL (100 μL/well) can bind Human TNFR1, His Tag (Cat. No. TN1-H5222) with a linear range of 3-25 ng/mL (QC tested).