Learn More
Sino Biological Recombinant Human TNFR-2 / CD120b Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 10417H08H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Asp257) of human TNFRII (NP_001057.1) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-211-6334