Learn More
Aviva Systems Biology Corp TNFRSF10B Antibody - N-terminal region (AVARP00032_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP00032_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TNFRSF10B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TNFRSF10B. Alternate names: CD262, DR5, KILLER, KILLER/DR5, TRAIL-R2, TRAILR2, TRICK2, TRICK2A, TRICK2B, TRICKB, ZTNFR9ApplicationWB. Species reactivity: Human
Catalog No. 50-014-62459