Learn More
Aviva Systems Biology Corp Tnfrsf12a Antibody - N-terminal region (ARP51328_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51328_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Tnfrsf12a. It was validated on Western Blot. . Alternate names: AI255180, C87282, Fn14, HPIP, TWEAK-R, TweakRApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-578