Learn More
Aviva Systems Biology Corp TNFRSF13B Antibody - N-terminal region (ARP63655_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63655_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TNFRSF13B. It was validated on Western Blot. . Alternate names: CD267, CVID, FLJ39942, MGC133214, MGC39952, TACI, TNFRSF14B, CVID2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-288-044