Learn More
Aviva Systems Biology Corp TNFRSF18 Antibody - C-terminal region (AVARP02052_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP02052_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TNFRSF18. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human TNFRSF18. Alternate names: AITR, GITR, CD357, GITR-DApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-62492