Learn More
Aviva Systems Biology Corp TNFRSF21 Antibody - N-terminal region (ARP47029_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47029_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TNFRSF21. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TNFRSF21. Alternate names: BM-018, DR6, MGC31965, CD358ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59454