Learn More
Aviva Systems Biology Corp TNFRSF25 Antibody - middle region (AVARP02066_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP02066_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TNFRSF25. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TNFRSF25. Alternate names: APO-3, DDR3, DR3, LARD, TNFRSF12, TR3, TRAMP, WSL-1, WSL-LRApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62501