Learn More
Cayman Chemical TNFRSF8CD30 LNg Isoform EX
SDP
Catalog No. 502949092
Encompass_Preferred
Source: Active recombinant C-terminal human IgG1 Fc-His-tagged CD30 expressed in HEK293 cells • Amino acids: 19-379 • MW: 66.5 kDa
Catalog No. 50-294-9092 Supplier Cayman Chemical Supplier No. 31829100 UG
Source: Active recombinant C-terminal human IgG1 Fc-His-tagged CD30 expressed in HEK293 cells • Amino acids: 19-379 • MW: 66.5 kDa