Learn More
Cayman Chemical TNFRSF8CD30C-Term RabbIt 100uL
SDP
Catalog No. 502913423
Encompass_Preferred
A BMI1 inhibitor inhibits proliferation of OVCAR-4 and OV-90 cells (IC50s 0 1 UM for both) induces BMI1 degradation in and apoptosis of the same cells reduces microtubule polymerization and induces cell cycle arrest at the G2/M phase in MDS-L cells at 3 and 0 03 UM respectively reduces tumor growth in a OV-90 orthotopic mouse model of ovarian cancer at 15 mg/kg biweekly
Catalog No. 50-291-3423 Supplier Cayman Chemical Supplier No. 35317100 UL
A BMI1 inhibitor inhibits proliferation of OVCAR-4 and OV-90 cells (IC50s 0 1 UM for both) induces BMI1 degradation in and apoptosis of the same cells reduces microtubule polymerization and induces cell cycle arrest at the G2/M phase in MDS-L cells at 3 and 0 03 UM respectively reduces tumor growth in a OV-90 orthotopic mouse model of ovarian cancer at 15 mg/kg biweekly