Learn More
Aviva Systems Biology Corp TNFRSF9 ANTIBODY 50UG/50UL
SDP
Supplier:  Aviva Systems Biology Corp ARP60755_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TNFRSF9. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human TNFRSF9. Alternate names: 4-1BB, CD137, CDw137, ILA, MGC2172ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rat
Catalog No. 50-287-148