Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents QP524450UG
Encompass_Preferred
Product Type: Recombinant Protein. Conjugate/Tag: Untagged. Host: E. coli. Species Reactivity: Human. BioActivity: Human TNFSF10 / TRAIL / APO-2L induces RPMI-8226 cell proliferation (typical ED50 is ? 15 ng/ml). AA Sequence: MRERGPQRVA AHITGTRGRS NTLSSPNSKN EKALGRKINS WESSRSGHSF LSNLHLRNGE LVIHEKGFYY IYSQTYFRFQ EEIKENTKND KQMVQYIYKY TSYPDPILLM KSARNSCWSK DAEYGLYSIY QGGIFELKEN DRIFVSVTNE HLIDMDHEAS FFGAFLVG. Buffer: 10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5. Purity: Greater than 90 as determined by SDS-PAGE. Endotoxin: Less than or equal to 1.00 EUs/ug as determined by the LAL method. Gene Symbol: TNFSF10. Gene ID: 8743. Uniprot: P50591.
Catalog No. 50-188-5733