Learn More
ACROBiosystems ACROBiosystems Human TNFSF11 / RANKL / CD254 Protein, His Tag (active trimer) (MALS verified)
Catalog No. 501957268
Encompass_Preferred
Recombinant Protein;1MG;Human TNFSF11, His Tag (active trimer) (MALS verified) (RAL-H5240) is expressed from human 293 cells (HEK293). It contains AA Gly 64 - Asp 245 (Accession # AAC51762.1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 22.4 kDa. The protein migrates as 30-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human RANK, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. RAK-H5251) at 0.5 μg/mL (100 μL/well) can bind Human TNFSF11, His Tag (active trimer) (MALS verified) (Cat. No. RAL-H5240) with a linear range of 0.4-6 ng/mL (QC tested).;Immobilized Human OPG, Fc Tag (Cat. No. TNB-H5259) at 2 μg/mL (100 μL/well) can bind Human TNFSF11, His Tag (active trimer) (MALS verified) (Cat. No. RAL-H5240) with a linear range of 0.2-2 ng/mL (Routinely tested).;Anti-TNFSF11 (human IgG1) captured on CM5 chip via Anti-human IgG Fc antibodies surface can bind Human TNFSF11, His Tag (active trimer) (MALS verified)
Catalog No. 50-195-7268 Supplier ACROBiosystems Supplier No. RALH52401MG
Recombinant Protein;1MG;Human TNFSF11, His Tag (active trimer) (MALS verified) (RAL-H5240) is expressed from human 293 cells (HEK293). It contains AA Gly 64 - Asp 245 (Accession # AAC51762.1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 22.4 kDa. The protein migrates as 30-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human RANK, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. RAK-H5251) at 0.5 μg/mL (100 μL/well) can bind Human TNFSF11, His Tag (active trimer) (MALS verified) (Cat. No. RAL-H5240) with a linear range of 0.4-6 ng/mL (QC tested).;Immobilized Human OPG, Fc Tag (Cat. No. TNB-H5259) at 2 μg/mL (100 μL/well) can bind Human TNFSF11, His Tag (active trimer) (MALS verified) (Cat. No. RAL-H5240) with a linear range of 0.2-2 ng/mL (Routinely tested).;Anti-TNFSF11 (human IgG1) captured on CM5 chip via Anti-human IgG Fc antibodies surface can bind Human TNFSF11, His Tag (active trimer) (MALS verified)